Learn More
RT1A Mouse anti-Rat, PE, Clone: OX-18, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559993
Description
The OX-18 antibody reacts with a non-polymorphic determinant of rat MHC class I antigen on erythrocytes and most nucleated cells, except normal neurons and glial cells. OX-18 mAb has been reported to block the recognition of RT1A by rat NK cells.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
RT1A | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
RT1B-depleted membrane glycoproteins isolated from Wistar rat spleen | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
OX-18 | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Rat | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.