Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTKN2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525716
Description
RTKN2 may play an important role in lymphopoiesis.
Specifications
RTKN2 | |
Polyclonal | |
Unconjugated | |
Q8IZC4, Q14B46 | |
RTKN2 | |
KLH conjugated synthetic peptide between 99-127 amino acids from the N-terminal region of human RTKN2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
RTKN2 | |
PLEKHK1, bA531F24.1, PH domain-containing family K member 1, pleckstrin homology domain containing, family K member 1, pleckstrin homology domain-containing family K member 1, rhotekin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170799, 219790 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction