Learn More
reticulon 2 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RTN2.
Supplier: Abnova Corporation H00006253A01
Description
This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: MGQVLPVFAHCKEAPSTASSTPDSTEGGNDDSDFRELHTAREFSEEDEEETTSQDWGTPRELTFSYIAFDGVVGSGGRRDSTARRPRPQGRSVSEPRDQHSpecifications
reticulon 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant RTN2. | |
RTN2 | |
NSP2/NSPL1 | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_005619 | |
RTN2 | |
RTN2 (NP_005610, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
6253 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.