Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubella Mouse anti-Human, Clone: EI-20, MilliporeSigma™

Catalog No. MAB925MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB925MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB925MI Supplier MilliporeSigma Supplier No. MAB925
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Rubella Antibody, E1, clone EI-20 is an antibody against Rubella for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rubella
Applications Immunofluorescence
Classification Monoclonal
Clone EI-20
Conjugate Unconjugated
Formulation Purified immunoglobulin presented as a liquid in 0.2Mphosphate buffer, 0.25MNaCl with 0.1% sodium azide.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.