Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubella Goat anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB1060MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1060MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1060MI Supplier MilliporeSigma Supplier No. AB1060
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Rubella using this Anti-Rubella Antibody validated for use in ELISA, immunofluoresence, western blot, HI.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rubella
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation IgG fraction (95% pure). In PBS (0.01 M, pH 7.2) with 0.1% sodium azide.
Host Species Goat
Immunogen Strain HPV77.
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.