Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUN domain containing 2A, Mouse, Clone: 6D6, Abnova™

Catalog No. 89001369 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-369 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-369 Supplier Abnova Corporation Supplier No. H00084127M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RUNDC2A.

Sequence: MDEERSSMLPTMAAGPNSILFAINIDNKDLNGQSKFAPTVSDLLKESTQNVTLLKESTQGVSSVFREITASSAISILIKPEQETDPLPVVSRNVSADAKCKKERKKKKQVTNIISFDDEEDEQNSGDMFKKTPGAGESSEDNSDHSSVNIMSAFESPFGPNSNGSQSSNSWKIDSLSLNREFGYQKLDVKSIDDEDVDENEDDVYGNSSGRKHRGHSESPEKNGAHSVTQAGVQWHDLGSLQPLPPGFR

Specifications

Antigen RUN domain containing 2A
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6D6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant RUNDC2A.
Formulation PBS with no preservative; pH 7.4
Gene RUNDC2A
Gene Accession No. BC041583
Gene Alias FLJ12363
Gene Symbols RUNDC2A
Host Species Mouse
Immunogen RUNDC2A (AAH41583, 1 a.a. ∼ 249 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84127
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.