Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX1, Mouse, Clone: 3A1, Abnova™

Catalog No. 89016450 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-450 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-450 Supplier Abnova Corporation Supplier No. H00000861M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RUNX1.

Core binding factor (CBF) is a heterodimeric transcription factor that binds to the core element of many enhancers and promoters. The protein encoded by this gene represents the alpha subunit of CBF and is thought to be involved in the development of normal hematopoiesis. Chromosomal translocations involving this gene are well-documented and have been associated with several types of leukemia. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: RVSPHHPAPTPNPRASLNHSTAFNPQPQSQMQDTRQIQPSPPWSYDQSYQYLGSIASPSVHPATPISPGRASGMTTLSAELSSRLSTAPDLTAFSDPRQFP

Specifications

Antigen RUNX1
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), In situ PLA, Western Blot
Classification Monoclonal
Clone 3A1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RUNX1.
Formulation PBS with no preservative; pH 7.4
Gene RUNX1
Gene Accession No. NM_001001890
Gene Alias AML1/AML1-EVI-1/AMLCR1/CBFA2/EVI-1/PEBP2aB
Gene Symbols RUNX1
Host Species Mouse
Immunogen RUNX1 (NP_001001890.1, 210 a.a. ∼ 310 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 861
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.