Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

runt-related transcription factor 3 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004049 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-049 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-049 Supplier Abnova Corporation Supplier No. H00000864A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RUNX3.

This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5'-PYGPYGGT-3' found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: FPDRFGDLERLRMRVTPSTPSPRGSLSTTSHFSSQPQTPIQGTSELNPFSDPRQFDRSFPTLPTLTESRFPDPRMHYPGAMSAAFP

Specifications

Antigen runt-related transcription factor 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RUNX3.
Formulation 50% glycerol
Gene RUNX3
Gene Accession No. NM_004350
Gene Alias AML2/CBFA3/FLJ34510/MGC16070/PEBP2aC
Gene Symbols RUNX3
Host Species Mouse
Immunogen RUNX3 (NP_004341, 194 a.a. ∼ 279 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 864
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.