Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Runx3 Mouse anti-Human, Clone: R3-5G4, MilliporeSigma™

Catalog No. MABE145MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE145MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE145MI Supplier MilliporeSigma Supplier No. MABE145
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-Runx3 Antibody, clone R3-5G4 (mouse monoclonal antibody) validated in western blot, immunoprecipitation to detect Runx3 also known as Runt-related transcription factor 3, Acute myeloid leukemia 2 protein, PEA2-alpha C, PEBP2-alpha C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Runx3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone R3-5G4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001026850
Gene Symbols RUNX3, AML2, CBFA3, PEBP2A3
Host Species Mouse
Immunogen Recombinant protein corresponding to human Runx3.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.