Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ryanodine Receptor Mouse anti-Bovine, Canine, Human, Mouse, Pig, Rat, Clone: RYR.1, MilliporeSigma™

Catalog No. MAB3086MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3086MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3086MI Supplier MilliporeSigma Supplier No. MAB3086
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Ryanodine Receptor Antibody, clone RYR.1 detects level of Ryanodine Receptor and has been published and validated for use in ELISA, flow cytometry, immunocytochemistry, immunohistochemistry, immunoprecipitation, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ryanodine Receptor
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone RYR.1
Conjugate Unconjugated
Formulation Purified immunoglobulin from culture supernatant. In 10mM sodium phosphate, 150mM NaCl, 0.01% NaN3, pH. 7.5
Gene Accession No. NM_000540.2; NM_001042723.1
Gene Symbols RYR1, CCO, RyR1, SKRR, RYR, MHS1, MHS, RYDR, CCD, RYR-1
Host Species Mouse
Immunogen Short synthetic polypeptide corresponding to the c-terminal domain of the ryanodine receptor.
Quantity 50 μg
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Pig, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.