Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-100 Protein Mouse anti-Human, Bovine, Mouse, Rat, Clone: 15E2E2, MilliporeSigma™
GSA_VA

Catalog No. MAB0791MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB0791MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB0791MI Supplier MilliporeSigma Supplier No. MAB0791
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-S-100 Protein Antibody, clone 15E2E2 is an antibody against S-100 Protein for use in immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen S-100 Protein
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 15E2E2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing PBS, pH 7.6, 1% BSA and 0.09% sodium azide.
Gene Accession No. NM_006271.1
Gene Symbols S100A1, S100A, S100, S100-alpha
Host Species Mouse
Immunogen Purified bovine S-100 protein
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Canine, Feline, Human, Mouse, Rabbit, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.