Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100 calcium binding protein A1, Mouse, Clone: 1D5, Abnova™

Catalog No. 89014181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-181 Supplier Abnova Corporation Supplier No. H00006271M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant S100A1.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-induced Ca2+ release, inhibition of microtubule assembly, and inhibition of protein kinase C-mediated phosphorylation. Reduced expression of this protein has been implicated in cardiomyopathies. [provided by RefSeq]

Sequence: MGSELETAMETLINVFHAHSGKEGDKYKLSKKELKELLQTELSGFLDAQKDVDAVDKVMKELDENGDGEVDFQEY

Specifications

Antigen S100 calcium binding protein A1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant S100A1.
Formulation PBS with no preservative; pH 7.4
Gene S100A1
Gene Accession No. NM_006271
Gene Alias S100/S100-alpha/S100A
Gene Symbols S100A1
Host Species Mouse
Immunogen S100A1 (NP_006262, 1 a.a. ∼ 75 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neurobiology
Primary or Secondary Primary
Gene ID (Entrez) 6271
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.