Learn More
Description
Specifications
Specifications
| Antigen | S100A12 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Liquid in PBS with 0.1% sodium azide. |
| Host Species | Rabbit |
| Immunogen | Full length recombinant His-tagged human S100A12 that was expressed in E. coli and purified by Ni-NTA affinity chromatography prior to immunization. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.