Learn More
S100 calcium binding protein A7, Mouse, Clone: 1C1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant S100A7.
Supplier: Abnova Corporation H00006278M03
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus. This protein is markedly over-expressed in the skin lesions of psoriatic patients, but is excluded as a candidate gene for familial psoriasis susceptibility. The exact function of this protein is not known. [provided by RefSeq
Sequence: MSNTQAERSIIGMIDMFHKYTRRDDKIDKPSLLTMMKENFPNFLSACDKKGTNYLADVFEKKDKNEDKKIDFSEFLSLLGDIATDYHKQSHGAAPCSGGSQSpecifications
S100 calcium binding protein A7 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC034687.1 | |
S100A7 | |
S100A7 (AAH34687.1, 1 a.a. ∼ 101 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
6278 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Immunofluorescence, Western Blot | |
1C1 | |
Mouse monoclonal antibody raised against a full length recombinant S100A7. | |
S100A7 | |
PSOR1/S100A7c | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.