Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A7 Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523142
Description
S100A7 (Psoriasin/HID-5) was originally identified as an up-regulated protein in psoriatic keratinocytes (1). Subsequently, the S100A7 protein was found to have elevated expression in abnormally differentiating primary keratinocytes and in various carcinomas, including ductal carcinoma in situ (2-3). S100A7 may play a role in the establishment or maintece of these pathologies, and may serve as a diagnostic marker of particular tumor types or abnormally proliferative epithelia.
Specifications
S100A7 | |
Polyclonal | |
Unconjugated | |
P31151 | |
S100A7 | |
Synthetic peptide (17 FHKYTRRDDKIDKPSL 32) of S100A7 protein. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Western Blot | |
0.5 mg/mL | |
S100A7 | |
PSOR1, S100A7c, S100 calcium-binding protein A7 (psoriasin 1), protein S100-A7, psoriasin 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6278 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction