Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Mouse anti-Human, Clone: 1F8, MilliporeSigma™

Catalog No. MABF296MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF296MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF296MI Supplier MilliporeSigma Supplier No. MABF296

Mouse Monoclonal Antibody

Anti-S100A8 Antibody, clone 1F8 (Azide Free) is an antibody against S100A8 for use in western blotting and ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A8
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F8
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.
Gene Accession No. NP_002955
Gene Symbols S100A8, CAGA, CFAG, MRP8
Host Species Mouse
Immunogen Recombinant protein corresponding to human S100A8.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.