Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8/MRP-8 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABF125MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF125MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF125MI Supplier MilliporeSigma Supplier No. ABF125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-S100A8/MRP-8 Antibody (Azide Free) is validated for use in Western Blotting and Immunohistochemistry and Activity Assay for the detection of S100A8/MRP-8 (Azide Free).
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A8/MRP-8
Applications Functional Assay, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum without preservatives.
Gene Symbols S100a8; Caga; Mrp8
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse S100A8/MRP-8.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_038678
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.