Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A9 Rat anti-Human, Mouse, Clone: 2A5, MilliporeSigma™

Catalog No. MABF290MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF290MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF290MI Supplier MilliporeSigma Supplier No. MABF290
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-S100A9 Antibody, clone 2A5 (Azide Free) is an antibody against S100A9 for use in western blotting and activity assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A9
Applications Functional Assay, Western Blot
Classification Monoclonal
Clone 2A5
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1 in buffer containing PBS without preservatives.
Gene Accession No. NP_00126878
Gene Symbols S100a9, Cagb, Mrp14
Host Species Rat
Immunogen Recombinant protein corresponding to mouse S100A9.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.