Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S6 (pS244) Mouse, Alexa Fluor 647, Clone: N5-676, BD

Catalog No. BDB560465 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560465 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560465 Supplier BD Biosciences Supplier No. 560465
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ribosomal protein S6 (∽29kDa calculated and ∽32kDa observed molecular weights) is a component of the 40S ribosomal subunit and belongs to the S6E family of ribosomal proteins. The S6 ribosomal protein plays a role in regulating the translation of RNAs and thus controlling the growth and proliferation of cells. S6 ribosomal protein phosphorylation, especially at multiple C-terminal serine residues S235, S236, S240, and S244, activates S6. The activated S6 ribosomal protein in turn upregulates the ribosomal translation of RNA species coding for other ribosomal proteins, peptide elongation factors and other proteins involved in cell cycle entry and progression. These phosphorylations are mediated by various kinases (e.g., p70S6K and PKCD) activated through cellular responses to growth factors, cytokines, tumor promoting agents, and mitogens. The S6 ribosomal protein can be dephosphorylated in growth-arrested cells. The N5-676 monoclonal antibody specifically detects the S6 ribosomal protein phosphorylated at S244.

Host Species: Mouse
Clone: N5-676
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human ribosomal protein S6 Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen S6 (pS244)
Applications Flow Cytometry
Classification Monoclonal
Clone N5-676
Conjugate Alexa Fluor 647
Description 40S ribosomal protein S6; Phosphoprotein NP33; RPS6; RS6
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human ribosomal protein S6 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.