Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

serum amyloid A4, constitutive, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018443 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-443 Supplier Abnova Corporation Supplier No. H00006291D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SAA4 protein.

Sequence: MRLFTGIVFCSLVMGVTSESWRSFFKEALQGVGDMGRAYWDIMISNHQNSNRYLYARGNYDAAQRGPGGVWAAKLISRSRVYLQGLIDYYLFGNSSTVLEDSKSNEKAEEWGRSGKDPDRFRPDGLPKKY

Specifications

Antigen serum amyloid A4, constitutive
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SAA4 protein.
Formulation PBS with no preservative; pH 7.4
Gene SAA4
Gene Accession No. NM_006512.1
Gene Alias C-SAA/CSAA
Gene Symbols SAA4
Host Species Rabbit
Immunogen SAA4 (NP_006503.1, 1 a.a. ∼ 130 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6291
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.