Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-SALL4 Clone: 6000,

Catalog No. DR1052100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DR1052100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DR1052100UG Supplier MilliporeSigma™ Supplier No. DR1052100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SALL4 Clone: 6000 in Human, Mouse samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immuno Blotting

This protein may be a zinc finger transcription factor. Defects in this gene are a cause of Duane-radial ray syndrome (DRRS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SALL4
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6000
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids 954-1054 of human SALL4, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽100kDa SALL4 protein in HeLa and NIH3T3 cells and human testes tissue.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.