Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SALL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 720030
Description
This antibody is predicted to react with Monkey, Rat, Mouse, Pig, Bovine, Rabbit and Dog. 720030 can be used for immunofluorescence analysis of Sall4 in human iPSCs and human ESC lines.
Sall4 protein is encoded by the Sall4 gene located on chromosome 20 in humans and belongs to the Spalt-like protein family. It plays an important role in somatic stem cell maintenance and is studied as a stem cell biomarker. It is a zinc finger transcription factor that is upregulated during development and shown to interact with Oct4 and Sox2. It also forms a complex with Nanog to maintain the pluripotency of Embryonic Stem cells (ESCs). Several current research studies focus on its function as an oncogene and tumor biomarker. Some of the reported target genes of Sall4 include p53, Bmi-1, Bcl2 and XIAP.
Specifications
SALL4 | |
Polyclonal | |
Unconjugated | |
SALL4 | |
5730441M18Rik; AA407717; AL022809; AW536104; C330011P20Rik; C78083; C78563; dJ1112F19.1; DRRS; HSAL4; SALL4; sal-like 4; sal-like 4 (Drosophila); sal-like protein 4; spalt like transcription factor 4; spalt-like transcription factor 4; Tex20; Zinc finger protein 797; zinc finger protein SALL4; ZNF797 | |
Rabbit | |
Affinity chromatography | |
RUO | |
57167, 99377 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
Q8BX22, Q9UJQ4 | |
SALL4 | |
Multiple peptides against Human Sall4 (aa 7-21, 1038-1053, 808-822). | |
100 μg | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction