Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salmonella Brazil Mouse anti-Human, Clone: 33/204, MilliporeSigma™

Catalog No. MAB748MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB748MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB748MI Supplier MilliporeSigma Supplier No. MAB748
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Salmonella Brazil Antibody, clone 33/204 is an antibody against Salmonella Brazil for use in ELISA and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Salmonella Brazil
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 33/204
Concentration 1 mg/1 mL
Conjugate Unconjugated
Formulation Purified immunoglobulin
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.