Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-SAMHD1 Clone: I19-18,

Catalog No. MABF933MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF933MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF933MI Supplier MilliporeSigma™ Supplier No. MABF933
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SAMHD1 Clone: I19-18 in Human samples, and it is validated for Flow Cytometry, Immunocytochemistry, Western Blotting

Deoxynucleoside triphosphate triphosphohydrolase SAMHD1 (UniProt Q9Y3Z3; also known as DCIP, Dendritic cell-derived IFNG-induced protein, dNTPase, Monocyte protein 5, MOP-5, SAM domain and HD domain-containing protein 1) is encoded by the SAMHD1 (also known as CHBL2, HDDC1, MOP5, SBBI88) gene (Gene ID 25939) in human. The sterile alpha motif domain and histidine-aspartate domain-containing protein 1 (SAMHD1) is a dGTP/GTP-activated deoxynucleotide (dNTP) triphosphohydrolase regulated by Cyclin A2/CDK1-dependent phosphorylation and is involved in balancing cellular dNTP pools. SAMHD1 was originally identified as a disease gene associated with the rare infantile encephalopathy Aicardi-Goutières syndrome. SAMHD1 was later shown to restrict HIV-1 replication in non-dividing myeloid-lineage cells and resting T cells by reducing the cellular dNTP concentration. Simian immunodeficiency viruses (SIVs) are found to use their Vpx/Vpr accessory proteins to engage and modify host Cullin-RING-type E3 ubiquitin ligases substrate recognition to target SAMHD1 for poly-ubiquitylation and subsequent proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAMHD1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone I19-18
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9Y3Z3
Gene Symbols SAMHD1, CHBL2, HDDC1, MOP5, SBBI88
Host Species Mouse
Immunogen GST-tagged recombinant full-length human SAMHD1.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_056289
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.