Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSAP Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514291
Description
PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.
Specifications
PSAP | |
Polyclonal | |
PSAP | |
Proactivator polypeptide; Saposin-A; Protein A; Saposin-B-Val; Saposin-B; Sphingolipid activator protein 1; SAP-1; Cerebroside sulfate activator; CSAct; Dispersin; Sulfatide/GM1 activator; Saposin-C; Co-beta-glucosidase; A1 activator; Glucosylceramidase activator; Sphingolipid activator protein 2; SAP-2; Saposin-D; Protein C; Component C; SAP1; GLBA; PSAP | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5660 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P07602 | |
PSAP | |
KLH conjugated synthetic peptide between 285-314 amino acids from the central region of human SAP | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction