Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAPAP1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABN309MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN309MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN309MI Supplier MilliporeSigma Supplier No. ABN309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This SAPAP1 antibody is validated for use in western blotting and immunohistochemistry for the detection of the SAPAP1 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPAP1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.
Gene Symbols DLGAP1; SAPAP1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human SAPAP1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.