Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-SAPK/JNK, Polyclonal

Catalog No. 559304200UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
55-930-4200UL 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-930-4200UL Supplier MilliporeSigma™ Supplier No. 559304200UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects SAPK/JNK Clone: in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry

The Stress-activated protein kinases (SAPK), also referred to as Jun N-terminal kinases (JNK), function in a protein kinase cascade transducing cellular stress signals. SAPK/JNKs are activated by a highly diverse group of extracellular signals, including UV light, inflammatory cytokines and a wide variety of cellular stresses. Activation occurs via phosphorylation at Thr183 and Tyr185 by the dual specificity enzyme SEK/MKK4 and as yet unidentified kinases. Since dual phosphorylation of SAPK/JNK at Thr183/Tyr185 is essential for kinase activity, phosphorylation at this site is an excellent marker of SAPK/JNK activity. Activated SAPK in turn phosphorylates and activates several transcription factors, including c-jun at Ser63/73 and ATF-2 at Ser69/71.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPK/JNK
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 10mM HEPES, 50% glycerol, 0.01% BSA, pH 7.5.
Host Species Rabbit
Immunogen a full-length, recombinant, human p54 SAPK/JNK2 fusion protein
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽54kDa SAPK/JNK protein in uv-treated HEK293 cells.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.