Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAT1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB11041622SS
Description
SAT1 Polyclonal specifically detects SAT1 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Goat, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SAT1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
DC21, diamine acetyltransferase 1, diamine N-acetyltransferase 1, EC 2.3.1.57, KFSD, Polyamine N-acetyltransferase 1, Putrescine acetyltransferase, SAT, Spermidine/spermine N(1)-acetyltransferase 1, spermidine/spermine N1-acetyltransferase, spermidine/spermine N1-acetyltransferase 1, spermidine/spermine N1-acetyltransferase alpha, SSAT-1, SSATKFSDX | |
Rabbit | |
27 kDa | |
0.025 mL | |
Cancer, Cell Biology, Hypoxia, Mitochondrial Markers | |
6303 | |
Human, Mouse, Rat, Porcine, Bovine, Chicken, Goat, Xenopus species | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
P21673 | |
SAT1 | |
A synthetic peptide made to a C-terminal region within residues 100-171 of human SAT1. [Swiss-Prot# P21673] | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat, pig, chicken, cow, goat, and Xenopus. Immunogen sequence has 86% homology to Zebrafish. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction