Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SBP-tag Mouse anti-Human, Clone: 20, MilliporeSigma™

Catalog No. MAB10764MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10764MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10764MI Supplier MilliporeSigma Supplier No. MAB10764
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SBP-tag Antibody, clone 20 detects level of SBP-tag and has been published and validated for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SBP-tag
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 20
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide
Gene Symbols SBC
Host Species Mouse
Immunogen Linear peptide corresponding to SBP tag.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.