Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAN domain containing 2 pseudogene, Mouse, Clone: 5F1, Abnova™

Catalog No. 89001300 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-300 Supplier Abnova Corporation Supplier No. H00054581M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SCAND2.

The SCAN domain is a highly conserved, leucine-rich motif of approximately 60 aa originally found within a subfamily of zinc finger proteins. This gene belongs to a family of genes that encode an isolated SCAN domain, but no zinc finger motif. Functional studies have established that the SCAN box is a protein interaction domain that mediates both hetero- and homoprotein associations, and maybe involved in regulation of transcriptional activity. Multiple transcript variants which encode the same isoform but differ only in their 3' UTRs, and another variant which encodes a distinct isoform have been described for this gene.

Sequence: MAVAVDQQIQTPSVQDLQIVKLEEDSHWEQEISLQGNYPGPETSCQSFWHFRYQEASRPREA

Specifications

Antigen SCAN domain containing 2 pseudogene
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5F1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SCAND2.
Formulation PBS with no preservative; pH 7.4
Gene SCAND2
Gene Accession No. NM_022050
Gene Symbols SCAND2
Host Species Mouse
Immunogen SCAND2 (NP_071333, 1 a.a. ∼ 62 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 54581
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.