Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

src kinase associated phosphoprotein 1, Mouse, Clone: 1C11, Abnova™

Catalog No. 89001094 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-094 Supplier Abnova Corporation Supplier No. H00008631M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SCAP1.

This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins. [provided by RefSeq

Sequence: MQAAALPEEIRWLLEDAEEFLAEGLRNENLSAVARDHRDHILRGFQQIKARYYWDFQPQGGDIGQDSSDDNHSGTLGLSLTSDAPFLSDYQDEGMEDIVK

Specifications

Antigen src kinase associated phosphoprotein 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SCAP1.
Formulation PBS with no preservative; pH 7.4
Gene SKAP1
Gene Accession No. NM_003726
Gene Alias SCAP1/SKAP55
Gene Symbols SKAP1
Host Species Mouse
Immunogen SCAP1 (NP_003717, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Receptors
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8631
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.