Learn More
src kinase associated phosphoprotein 2 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant SCAP2.
Supplier: Abnova Corporation H00008935A01
Description
The protein encoded by this gene belongs to the src family kinases. This protein is similar to the src kinase associated phosphoprotein 1. It is an adaptor protein that is thought to play an essential role in the src signaling pathway in various cells. It inhibits PTK2B/RAFTK activity and regulates alpha-synuclein phosphorylation. [provided by RefSeq
Sequence: MPNPSSTSSPYPLPEEIRNLLADVETFVADILKGENLSKKAKEKRESLIKKIKDVKSIYLQEFQDKGDAEDGEEYDDPFAGPPDTISLASERYDKDDEAPSDGAQFPPIAAQDLPFVLKAGYLEKRRKDHSFLGFEWQKRWCALSKTVFYYYGSDKDKQQKGEFAIDGYSVRMNNTLRKDGKKDCCFEISAPDKRIYQFTAASPKDAEEWVQQLKFVLQDMESDIIPEDYDERGELYDDVDHPLPISNPLTSSQPIDDEIYEELPEEEEDSAPVKVEEQRKMSQDSVHHTSGDKSTDYANFYQGLWDCTGAFSDELSFKRGDVIYILSKEYNRYGWWVGEMKGAIGLVPKAYIMEMYDISpecifications
src kinase associated phosphoprotein 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant SCAP2. | |
SKAP2 | |
MGC10411/MGC33304/PRAP/RA70/SAPS/SCAP2/SKAP-HOM/SKAP55R | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC002893 | |
SKAP2 | |
SCAP2 (AAH02893, 1 a.a. ∼ 359 a.a) full-length recombinant protein with GST tag. | |
RUO | |
Yes | |
8935 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.