Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCARB2, Mouse, Clone: 1C8, Abnova™

Catalog No. 89016498 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-498 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-498 Supplier Abnova Corporation Supplier No. H00000950M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SCARB2.

The protein encoded by this gene is a type III glycoprotein that is located primarily in limiting membranes of lysosomes and endosomes. Studies of the similar protein in mice and rat suggested that this protein may participate in membrane transportation and the reorganization of endosomal/lysosomal compartment. Deficiency of the similar protein in mice was reported to impair cell membrane transport processes and cause pelvic junction obstruction, deafness, and peripheral neuropathy. [provided by RefSeq

Sequence: FPHFYQADERFVSAIEGMHPNQEDHETFVDINPLTGIILKAAKRFQINIYVKKLDDFVETGDIRTMVFPVMYLNESVHIDKETASRLKSMINTTLIITN

Specifications

Antigen SCARB2
Applications ELISA
Classification Monoclonal
Clone 1C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SCARB2.
Formulation PBS with no preservative; pH 7.4
Gene SCARB2
Gene Accession No. NM_005506
Gene Alias AMRF/CD36L2/HLGP85/LIMPII/SR-BII
Gene Symbols SCARB2
Host Species Mouse
Immunogen SCARB2 (NP_005497, 339 a.a. ∼ 437 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 950
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.