Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

secretogranin II (chromogranin C), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89018853 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-853 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-853 Supplier Abnova Corporation Supplier No. H00007857A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SCG2.

The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Studies in rodents suggest that the full-length protein, secretogranin II, is involved in the packaging or sorting of peptide hormones and neuropeptides into secretory vesicles. The full-length protein is cleaved to produce the active peptide secretoneurin, which exerts chemotaxic effects on specific cell types, and EM66, whose function is unknown. [provided by RefSeq

Sequence: LARMLVKYPEIINSNQVKRVPGQGSSEDDLQEEEQIEQAIKEHLNQGSSQETDKLAPVSKRFPVGPPKNDDTPNRQYWDEDLLMKVLEYLNQEKAEKGREHIAKRAMENM

Specifications

Antigen secretogranin II (chromogranin C)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SCG2.
Formulation 50% glycerol
Gene SCG2
Gene Accession No. NM_003469
Gene Alias CHGC/SN/SgII
Gene Symbols SCG2
Host Species Mouse
Immunogen SCG2 (NP_003460, 508 a.a. ∼ 617 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 7857
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.