Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scirr1 Rabbit anti-Rat, Mouse, Bovine, Canine, Horse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB10223
Description
This Anti-Scirr1 Antibody is validated for use in western blotting for the detection of Scirr1.
Specifications
Scirr1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Bovine, Canine, Equine, Mouse, Rat |
Western Blot | |
Unconjugated | |
Scirr1; FBXL16; C16orf22; FBL16; Fbl16 | |
KLH-conjugated linear peptide corresponding to the internal domain of rat Scirr1. | |
100 μL | |
NP_001009504 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction