Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Sclerostin Clone: 7B6.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS445
Description
Specifically detects Sclerostin Clone: 7B6.1 in Human, Rat samples, and it is validated for Immunohistochemistry, Western Blotting
Sclerostin (UniProt Q9BQB4) is encoded by the SOST (also known as CDD, SOST1, VBCH) gene (ORP UNQ2976/PRO7455/PRO7476; Gene ID 50964) in human. Sclerostin is a glycoprotein expressed in osteocytes and functions as a negatively regulator against bone mass by antagonizing low density lipoprotein receptor-related proteins- (LRP5-/LRP6-) mediated WNT/beta-catenin pathway signaling. Sclerostin levels are increased in the post-menopausal women and elderly and in long term immobilized patients. Sclerostin neutralizing antibody is reported to enhance bone formation, mass, and strength in aged male rats. Sclerostin and its inhibitors have potential in prosthetic, regenerative and preventive therapy in dentistry.
Specifications
Sclerostin | |
Monoclonal | |
Unconjugated | |
Q9BQB4 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
Detects Sclerostin isoform 1 (UniProt Q9BQB4-1). Reactivity toward the spliced isoform 2 (UniProt Q9BQB4-2) is likely, but has not been determined. | |
-20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles | |
IgG2b κ |
Immunohistochemistry, Western Blot | |
7B6.1 | |
Mouse monoclonal IgG2bκ antibody tissue culture supernatant without preservatives. | |
SOST | |
GST-tagged recombinant protein corresponding to the internal fragment of human Sclerostin. | |
200 μL | |
Signaling | |
NP_079513 | |
Human, Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction