Learn More
SCN8A Sodium Channel Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | SCN8A Sodium Channel |
| Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.025% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 |
| Gene Symbols | SCN8A; Nav1.6; MED; NaCh6; PN4; CerIII |
| Host Species | Rabbit |
| Immunogen | Peptide corresponding to amino acids 1042-1061 from mouse (Accession number AAD20438) or rat (Accession number AAC26014) Scn8a. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.