Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDF-1 Mouse anti-Human, Mouse, Clone: K15C, MilliporeSigma™

Catalog No. MABC184MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC184MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC184MI Supplier MilliporeSigma Supplier No. MABC184
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect SDF using this mouse monoclonal antibody, Anti-SDF-1 Antibody, clone K15C, Azide free validated for use in western blotting, immunohistochemistry, Neutralizing and ELISA.

Specifications

Antigen SDF-1
Applications ELISA, Immunohistochemistry, Neutralization, Western Blot
Classification Monoclonal
Clone K15C
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing PBS without preservatives.
Gene Accession No. NP_000600.1
Gene Symbols CXCL12, SDF1, SDF1A, SDF1B
Host Species Mouse
Immunogen BSA-conjugated linear peptide corresponding to human SDF-1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.