Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-SDHB Clone: 10C5.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABN627
Description
Specifically detects SDHB Clone: 10C5.1 in Human, Mouse samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
Succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial (EC 1.3.5.1; UniProt P21912; also known as Ip, Iron-sulfur subunit of complex II) is encoded by the SDHB (also known as IP, SDH, CWS2, PGL4, SDH1, SDHIP) gene (Gene ID 6390) in human. Originally described by Otto Warburg and now commonly known as the Warburg effect refers to the phenomenon that many tumor cells use aerobic glycolysis instead of mitochondrial respiration for energy production to support cell growth and proliferation. Enzymes involved in the complex network of metabolic pathways are therefore being studied as potential cancer markers and targets for treatment. Succinate dehydrogenase (SDH), also known as mitochondrial respiratory Complex II, is a holoenzyme consisting of four essential subunits, SDHA (a flavoprotein), SDHB (iron-sulfur protein), and two membrane anchor units SDHC and SDHD. SDH assembly requires two factors, SDHAF1 and SDHAF2, while its function is influenced by the deacetylase activity of SIRT3. SDH is the only enzyme complex known to participate in both the TCA cycle and electron transport chain (ETC.). SDH catalyzes the oxidation of succinate in the TCA to form fumarate, a reaction that is coupled to the reduction of ubiquinone to ubiquinol in the ETC. SDH loss-of-function mutations have been linked to pheochromocytoma (PCC), paraganglioma (PGL), gastrointestinal stromal tumor (GIST), renal cell carcinoma, and ovarian cancer.
Specifications
SDHB | |
Monoclonal | |
Unconjugated | |
P21912 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Immunohistochemistry (Paraffin), Western Blot | |
10C5.1 | |
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
SDHB, SDH, SDH1 | |
GST-tagged recombinant protein corresponding to the C-terminal half of human SDHB. | |
100 μg | |
Neuroscience | |
NP_002991 | |
2°C to 8°C for one year from date of shipment | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction