Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC24D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220280
Description
SEC24D Polyclonal specifically detects SEC24D in Human samples. It is validated for Western Blot.Specifications
SEC24D | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
FLJ43974, KIAA0755member D, SEC24 family, member D (S. cerevisiae), SEC24 related gene family, member D | |
Rabbit | |
113 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (89%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1.01mg/mL | |
Western Blot 1:500-1:3000 | |
O94855 | |
SEC24D | |
Recombinant protein encompassing a sequence within the C-terminus region of human SEC24D. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
9871 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction