Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sec31A Mouse anti-Human, Unlabeled, Clone: 32, BD

Catalog No. BDB612351 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612351 150 μg
BDB612350 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612351 Supplier BD Biosciences Supplier No. 612351
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Eukaryotic protein trafficking involves packaging of target molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their contents into an acceptor compartment. Vesicles that bud from the Golgi cisternae and from the ER contain a non-clathrin based coat. This coat is an oligomeric complex whose subunits are termed COPs (coatomer proteins). COPI is the golgi- and endosome-associated COP complex, while COPII is the ER-associated coat complex. In yeast, the COPII complex has a cargo binding subcomplex that includes Sar1p, Sec23p, and Sec24p, which recruits Sec13p and Sec31p leading to vesicle formation. The human homologues of Sec31p are Sec31A and Sec31B. Sec31A has 40% identity with Sec31B, and contains an N-terminal WD-40 domain and a C-terminal proline-rich region. Sec31A mRNA is widely expressed, while Sec31B is found only in testis and thymus. In HeLa, Sec31A localizes to vesicular structures in the perinuclear region of the cell, and co-localizes with the COPII component Sec13. Antibodies against Sec31A inhibit ER to Golgi transport of vesicular stomatitis G protein. Thus, Sec31 may be an important COPII component involved in ER to Golgi transport, as well as a marker for COPII vesicles.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sec31A
Applications Western Blot
Classification Monoclonal
Clone 32
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Sec31A aa. 522-719
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.