Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

selenoprotein I, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-753 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-753 Supplier Abnova Corporation Supplier No. H00085465A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SELI.

This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. [provided by RefSeq]

Sequence: MAGYEYVSPEQLAGFDKYKYSAVDTNPLSLYVMHPFWNTIVKVFPTWLAP

Specifications

Antigen selenoprotein I
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SELI.
Formulation 50% glycerol
Gene SELI
Gene Accession No. NM_033505
Gene Alias KIAA1724
Gene Symbols SELI
Host Species Mouse
Immunogen SELI (NP_277040, 1 a.a. ∼ 50 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 85465
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.