Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

selectin P (granule membrane protein 140kDa, antigen CD62), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018263 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-263 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-263 Supplier Abnova Corporation Supplier No. H00006403D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SELP protein.

This gene encodes a 140kDa protein that is stored in the alpha-granules of platelets and Weibel-Palade bodies of endothelial cells. This protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. The membrane protein is a calcium-dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens on neutrophils and monocytes. Alternative splice variants may occur but are not well documented. [provided by RefSeq

Sequence: MANCQIAILYQRFQRVVFGISQLLCFSALISELTNQKEVAAWTYHYSTKAYSWNISRKYCQNRYTDLVAIQNKNEIDYLNKVLPYYSSYYWIGIRKNNKTWTWVGTKKALTNEAENWADNEPNNKRNNEDCVEIYIKSPSAPGKWNDEHCLKKKHALCYTASCQDMSCSKQGECLETIGNYTCSCYPGFYGPECEYVRECGELELPQHVLMNCSHPLGNFSFNSQCSFHCTDGYQVNGPSKLECLASGIWTNKPPQCLAAQCPPLKIPERGNMTCLHSAKAFQHQSSCSFSCEEGFALVGPEVVQCTASGVWTAPAPVCKAVQCQHLEAPSEGTMDCVHPLTAFAYGSSCKFECQPGYRVRGLDMLRCIDSGHWSAPLPTCEAISCEPLESPVHGSMDCSPSLRAFQYDTNCSFRCAEGFMLRGADIVRCDNLGQWTAPAPVCQALQCQDLPVPNEARVNCSHPFGAFRYQSVCSFTCNEGLLLVGASVLQCLATGNWNSVPPECQAIPCTPLLSPQNGTMTCVRPLGSSSYKSTCQFICDEGYSLSGPERLDCTRSGRWTDSPPMCEAIKCPELFAPEQGSLDCSDTRGEFNVGSTCHFSCNNGFKLEGPNNVECTTSGRWSATPPTCKGIASLPTPGVQCPALTTPGQGTMYCRHHPGTFGFNTTCYFGCNAGFTLIGDSTLSCRPSGQWTAVTPACRAVKCSELHVNKPIAMNCSNLWGNFSYGSICSFHCLEGQLLNGSAQTACQENGHWSTTVPTCQGPLTIQEALTYFGGAVASTIGLIMGGTLLALLRKRFRQKDDGKCPLNPHSHLGTYGVFTNAAFDPSP

Specifications

Antigen selectin P (granule membrane protein 140kDa, antigen CD62)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SELP protein.
Formulation PBS with no preservative; pH 7.4
Gene SELP
Gene Accession No. BC068533
Gene Alias CD62/CD62P/FLJ45155/GMP140/GRMP/LECAM3/PADGEM/PSEL
Gene Symbols SELP
Host Species Rabbit
Immunogen SELP (AAH68533.1, 1 a.a. ∼ 829 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Gene ID (Entrez) 6403
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.