Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA4A Mouse anti-Human, PE, Clone: T9-10, BD

Catalog No. BDB564812 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564812 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564812 Supplier BD Biosciences Supplier No. 564812

Mouse Monoclonal Antibody

The T9-10 monoclonal antibody specifically binds to Semaphorin 4A (SEMA4A/ SEM4A) which is also known as Semaphorin-B (Sema B/ SEMAB/SEMB). SEMA4A is a type I transmembrane glycoprotein that belongs to the class IV subfamily of the semaphorin family. A cleaved, soluble form of SEMA4A has also been described. It is expressed by B cells, dendritic cells, monocytes, granulocytes, and at low levels by some T cells. SEMA4A plays several roles in the immune system including the reciprocal activation of antigen presenting cells and T cells leading to the differentiation of effector CD4+ T cells. SEMA4A functions in the developing nervous system by guiding neuronal axons to their proper target cells. It is also involved in the regulation of angiogenesis and the progression of certain cancers.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SEMA4A
Applications Flow Cytometry
Classification Monoclonal
Clone T9-10
Concentration 0.2mg/mL
Conjugate PE
Description Semaphorin-4A; SEMA4A; SEM4A; Semaphorin-B; Sema B; SEMB; SEMAB; CORD10
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human Semaphorin 4A Protein
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.