Learn More
SEMA4G, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SEMA4G.
Supplier: Abnova Corporation H00057715A01
Description
Semaphorins are a large family of conserved secreted and membrane associated proteins which possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Based on sequence and structural similarities, semaphorins are put into eight classes: invertebrates contain classes 1 and 2, viruses have class V, and vertebrates contain classes 3-7. Semaphorins serve as axon guidance ligands via multimeric receptor complexes, some (if not all) containing plexin proteins. This gene encodes a class 4 semaphorin. This gene and the gene for mitochondrial ribosomal protein L43 overlap at map location 10q24.31 and are transcribed in opposite directions. [provided by RefSeq
Sequence: RRPSRELDATPRMTIPYEELSGTRHFKGQAQNYSTLLLEEASARLLVGARGALFSLSANDIGDGAHKEIHWEASPEMQSKCHQKGKNNQTECSpecifications
SEMA4G | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SEMA4G. | |
SEMA4G | |
FLJ20590/KIAA1619/MGC102867 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_017893 | |
SEMA4G | |
SEMA4G (NP_060363, 24 a.a. ∼ 115 a.a) partial recombinant protein with GST tag. | |
RUO | |
57715 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.