Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA4G, Mouse, Clone: 3F2, Abnova™

Catalog No. 89002650 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-650 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-650 Supplier Abnova Corporation Supplier No. H00057715M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SEMA4G.

Semaphorins are a large family of conserved secreted and membrane associated proteins which possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Based on sequence and structural similarities, semaphorins are put into eight classes: invertebrates contain classes 1 and 2, viruses have class V, and vertebrates contain classes 3-7. Semaphorins serve as axon guidance ligands via multimeric receptor complexes, some (if not all) containing plexin proteins. This gene encodes a class 4 semaphorin. This gene and the gene for mitochondrial ribosomal protein L43 overlap at map location 10q24.31 and are transcribed in opposite directions. [provided by RefSeq

Sequence: RRPSRELDATPRMTIPYEELSGTRHFKGQAQNYSTLLLEEASARLLVGARGALFSLSANDIGDGAHKEIHWEASPEMQSKCHQKGKNNQTEC

Specifications

Antigen SEMA4G
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SEMA4G.
Formulation PBS with no preservative; pH 7.4
Gene SEMA4G
Gene Accession No. NM_017893
Gene Alias FLJ20590/KIAA1619/MGC102867
Gene Symbols SEMA4G
Host Species Mouse
Immunogen SEMA4G (NP_060363, 24 a.a. ∼ 115 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57715
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.