Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018708 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-708 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-708 Supplier Abnova Corporation Supplier No. H00057556D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SEMA6A protein.

Sequence: MRSEALLLYFTLLHFAGAGFPEDSEPISISHGNYTKQYPVFVGHKPGRNTTQRHRLDIQMIMIMNGTLYIAARDHIYTVDIDTSHTEEIYCSKKLTWKSRQADVDTCRMKGKHKDECHNFIKVLLKKNDDALFVCGTNAFNPSCRNYKMDTLEPFGDEFSGMARCPYDAKHANVALFADGKLYSATVTDFLAIDAVIYRSLGESPTLRTVKHDSKWLKEPYFVQAVDYGDYIYFFFREIAVEYNTMGKVVFPRVAQVCKNDMGGSQRVLEKQWTSFLKARLNCSVPGDSHFYFNILQAVTDVIRINGRDVVLATFSTPYNSIPGSAVCAYDMLDIASVFTGRFKEQKSPDSTWTPVPDERVPKPRPGCCAGSSSLERYATSNEFPDDTLNFIKTHPLMDEAVPSIFNRPWFLRTMVRYRLTKIAVDTAAGPYQNHTVVFLGSEKGIILKFLARIGNSGFLNDSLFLEEMSVYNSEKCSYDGVEDKRIMGMQLDRASSSLYVAFSTCVIKVPLGRCERHGKCKKTCIASRDPYCGWIKEGGACSHLSPNSRLTFEQDIERGNTDGLGDCHNSFVALNGHSSSLLPSTTTSDSTAQEGYESRGGMLDWKHLLDSPDSTDPLGAVSSHNHQDKKGVIRESYLKGHDQLVPVTLLAIAVILAFVMGAVFSGITVYCVCDHRRKDVAVVQRKEKELTHSRRGSMSSVTKLSGLFGDTQSKDPKPEAILTPLMHNGKLATPGNTAKMLIKADQHHLDLTALPTPESTPTLQQKRKPSRGSREWERNQNLINACTKDMPPMGSPVIPTDLPLRASPSHIPSVVVLPITQQGYQHEYVDQPKMSEVAQMALEDQAATLEYKTIKEHLSSKSPNHGVNLVENLDSLPPKVPQREASLGPPGASLSQTGLSKRLEMHHSSSYGVDYKRSYPTNSLTRSHQATTLKRNNTNSSNSSHLSRNQSFGRGDNPPPAPQRVDSIQVHSSQPSGQAVTVSRQPSLNAYNSLTRSGLKRTPSLKPDVPPKPSFAPLSTSMKPNDACT

Specifications

Antigen sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SEMA6A protein.
Formulation PBS with no preservative; pH 7.4
Gene SEMA6A
Gene Accession No. NM_020796
Gene Alias HT018/KIAA1368/SEMA/SEMA6A1/SEMAQ/VIA
Gene Symbols SEMA6A
Host Species Rabbit
Immunogen SEMA6A (NP_065847.1, 1 a.a. ∼ 1030 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 57556
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.