Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA6D, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89003203 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-203 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-203 Supplier Abnova Corporation Supplier No. H00080031D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SEMA6D protein.

Semaphorins are a large family, including both secreted and membrane associated proteins, many of which have been implicated as inhibitors or chemorepellents in axon pathfinding, fasciculation and branching, and target selection. All semaphorins possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Additional sequence motifs C-terminal to the semaphorin domain allow classification into distinct subfamilies. Results demonstrate that transmembrane semaphorins, like the secreted ones, can act as repulsive axon guidance cues. This gene encodes a class 6 vertebrate transmembrane semaphorin that demonstrates alternative splicing. Six transcript variants have been identified and expression of the distinct encoded isoforms is thought to be regulated in a tissue- and development-dependent manner. [provided by RefSeq

Sequence: MRVFLLCAYILLLMVSQLRAVSFPEDDEPLNTVDYHYSRQYPVFRGRPSGNESQHRLDFQLMLKIRDTLYIAGRDQVYTVNLNEMPKTEVIPNKKLTWRSRQQDRENCAMKGKHKDECHNFIKVFVPRNDEMVFVCGTNAFNPMCRYYRLSTLEYDGEEISGLARCPFDARQTNVALFADGKLYSATVADFLASDAVIYRSMGDGSALRTIKYDSKWIKEPHFLHAIEYGNYVYFFFREIAVEHNNLGKAVYSRVARICKNDMGGSQRVLEKHWTSFLKARLNCSVPGDSFFYFDVLQSITDIIQINGIPTVVGVFTTQLNSIPGSAVCAFSMDDIEKVFKGRFKEQKTPDSVWTAVPEDKVPKPRPGCCAKHGLAEAYKTSIDFPDETLSFIKSHPLMDSAVPPIADEPWFTKTRVRYRLTAISVDHSAGPYQNYTVIFVGSEAGMVLKVLAKTSPFSLNDSVLLEEIEAYNHAK

Specifications

Antigen SEMA6D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SEMA6D protein.
Formulation PBS with no preservative; pH 7.4
Gene SEMA6D
Gene Accession No. NM_024966.2
Gene Alias FLJ11598/KIAA1479
Gene Symbols SEMA6D
Host Species Rabbit
Immunogen SEMA6D (NP_079242.2, 1 a.a. ∼ 476 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80031
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.