Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA7A, Mouse, Clone: 1G1, Abnova™

Catalog No. 89002729 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-729 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-729 Supplier Abnova Corporation Supplier No. H00008482M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SEMA7A.

The protein encoded by this gene binds to cell surfaces through a glycosylphosphatidylinositol (GPI) linkage. The encoded glycoprotein is found on activated lymphocytes and erythrocytes. This protein may be involved in immunomodulatory and neuronal processes. Defects in this gene can result in loss of bone mineral density (BMD). Three transcript variants encoding different isoforms have been found for this gene

Sequence: EPHKECPNPKPDKAPLQKVSLAPNSRYYLSCPMESRHATYSWRHKENVEQSCEPGHQSPNCILFIENLTAQQYGHYFCEAQEGSYFREAQHWQLLPED

Specifications

Antigen SEMA7A,
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1G1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SEMA7A.
Formulation PBS with no preservative; pH 7.4
Gene SEMA7A
Gene Accession No. NM_003612
Gene Alias CD108/CDw108/H-SEMA-K1/H-Sema-L/JMH/MGC126692/MGC126696/SEMAK1/SEMAL
Gene Symbols SEMA7A
Host Species Mouse
Immunogen SEMA7A (NP_003603, 536 a.a. ∼ 633 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8482
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.