Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Semaphorin 3A, APC, Clone: 215803, R&D Systems™

Catalog No. IC1250A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1250A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1250A Supplier R&D Systems Supplier No. IC1250A
This item is not returnable. View return policy

Applications:

Flow Cytometry (Intracellular Staining)

Specifications

Antigen Semaphorin 3A
Applications Flow Cytometry
Classification Monoclonal
Clone 215803
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q14563
Gene Alias (semaphorin) 3A, coll-1, collapsin 1, Hsema-I, Hsema-III, MGC133243, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema III, SEMA1, Sema3A, SEMAD, SEMAIII, SEMAL, semaphorin D, Semaphorin III, semaphorin L, semaphorin-3A, SemD
Gene Symbols SEMA3A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3A, Lys26-Val771 (Arg555Ala, Arg552Ala), Accession # Q14563
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10371
Test Specificity Detects human Semaphorin 3A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Semaphorin 3B, 3E, 6A, 6B, 6C, 6D, 7A, recombinant mouse Semaphorin 3C, or 3F is observed.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.